siRNA Details
| Gene Symbol: Pus10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025278.1 | NP_001020449.1 | 9 | 1737 | ||
| GenBank mRNA | BC093611.1 | - | 9 | 1737 | 7307810 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pus10| Gene Symbol: Pus10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025278.1 | NP_001020449.1 | 9 | 1737 | ||
| GenBank mRNA | BC093611.1 | - | 9 | 1737 | 7307810 | |