siRNA Details
| Gene Symbol: Reep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025279.1 | NP_001020450.1 | 6 | 488 | ||
| GenBank mRNA | BC097957.1 | - | 6 | 488 | 7462731 | |
| FQ234004.1 | - | 7 | 715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reep4| Gene Symbol: Reep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025279.1 | NP_001020450.1 | 6 | 488 | ||
| GenBank mRNA | BC097957.1 | - | 6 | 488 | 7462731 | |
| FQ234004.1 | - | 7 | 715 | |||