siRNA Details
| Gene Symbol: Fam118b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025283.1 | NP_001020454.1 | 5 | 814 | ||
| XM_006242784.2 | XP_006242846.1 | 6 | 894 | |||
| XM_008766056.1 | XP_008764278.1 | 6 | 984 | |||
| GenBank mRNA | BC098017.1 | - | 5 | 814 | 7454942 | |

Fam118b
Genomic Map