siRNA Details
| Gene Symbol: DPPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025290.2 | NP_001020461.1 | 3 | 426 | ||
| GenBank mRNA | BX092581.1 | - | 3 | 381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPPA5| Gene Symbol: DPPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025290.2 | NP_001020461.1 | 3 | 426 | ||
| GenBank mRNA | BX092581.1 | - | 3 | 381 | ||