siRNA Details
| Gene Symbol: HMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005519.1 | NP_005510.1 | 2 | 1139 | ||
| XM_005269743.3 | XP_005269800.1 | 3 | 1001 | |||
| GenBank mRNA | BC132758.1 | - | 2 | 949 | 40146780 | |
| BC137139.1 | - | 2 | 949 | 9021136 | ||

HMX2
Genomic Map