siRNA Details
| Gene Symbol: PMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199653.1 | NP_001186582.1 | 5 | 672 | ||
| NM_001199654.1 | NP_001186583.1 | 5 | 739 | |||
| NM_007221.3 | NP_009152.2 | 5 | 733 | |||
| GenBank mRNA | AF141310.1 | - | 5 | 696 | ||
| AK124646.1 | - | 5 | 667 | |||
| AK289490.1 | - | 5 | 714 | |||
| AK290260.1 | - | 5 | 714 | |||
| AL080101.1 | - | 5 | 702 | |||
| BC050735.1 | - | 5 | 688 | |||
| BC056417.1 | - | 5 | 697 | |||
| BC065031.1 | - | 5 | 712 | 6148250 | ||

PMF1
Genomic Map