siRNA Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031765.1 | NP_113953.1 | 9 | 1411 | ||
| XM_006250212.2 | XP_006250274.1 | 8 | 1555 | |||
| GenBank mRNA | AJ223083.1 | - | 8 | 946 | ||
| BC091363.1 | - | 9 | 1411 | 7319081 | ||

Rxrg
Genomic Map