siRNA Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031765.1 | NP_113953.1 | 10 | 1632 | ||
| XM_006250212.2 | XP_006250274.1 | 9 | 1776 | |||
| GenBank mRNA | BC091363.1 | - | 10 | 1632 | 7319081 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxrg| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031765.1 | NP_113953.1 | 10 | 1632 | ||
| XM_006250212.2 | XP_006250274.1 | 9 | 1776 | |||
| GenBank mRNA | BC091363.1 | - | 10 | 1632 | 7319081 | |