siRNA Details
| Gene Symbol: ZMAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011657.3 | NP_001011657.2 | 7 | 3420 | ||
| NM_001282400.1 | NP_001269329.1 | 10 | 3976 | |||
| NM_001282401.1 | NP_001269330.1 | 9 | 3923 | |||
| XM_005262212.4 | XP_005262269.2 | 7 | 3437 | |||
| XM_005262213.4 | XP_005262270.2 | 7 | 3434 | |||
| XM_005262214.3 | XP_005262271.2 | 6 | 3368 | |||
| XM_005262216.4 | XP_005262273.2 | 7 | 3139 | |||
| XM_006724711.3 | XP_006724774.1 | 3 | 2899 | |||
| XM_017029903.1 | XP_016885392.1 | 7 | 3136 | |||
| XM_017029904.1 | XP_016885393.1 | 7 | 5189 | |||

ZMAT1
Genomic Map