siRNA Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032532.2 | NP_115921.2 | 11 | 4158 | ||
| XM_011536190.2 | XP_011534492.1 | 10 | 4081 | |||
| XM_011536191.2 | XP_011534493.1 | 8 | 3799 | |||
| GenBank mRNA | AB058769.2 | - | 9 | 3644 | ||
| AJ586132.1 | - | 1 | 78 | |||
| AK127635.1 | - | 1 | 166 | |||
| AL832410.1 | - | 1 | 375 | |||
| BC146783.1 | - | 9 | 3644 | 9007239 | ||
| BC150607.1 | - | 10 | 3833 | 9056977 | ||
| DQ009660.1 | - | 11 | 4158 | |||

FNDC1
Genomic Map