siRNA Details
| Gene Symbol: ZNF628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033113.2 | NP_149104.3 | 3 | 3833 | ||
| XM_005259371.3 | XP_005259428.1 | 2 | 3478 | |||
| GenBank mRNA | BC047332.1 | - | 1 | 991 | ||
| BC089449.1 | - | 3 | 3833 | 30560806 | ||

ZNF628
Genomic Map