siRNA Details
| Gene Symbol: ZNF518B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053042.2 | NP_444270.2 | 3 | 2627 | ||
| XM_005248193.2 | XP_005248250.1 | 2 | 2462 | |||
| XM_017008784.1 | XP_016864273.1 | 2 | 5085 | |||
| XM_017008785.1 | XP_016864274.1 | 3 | 2853 | |||
| XM_017008786.1 | XP_016864275.1 | 2 | 2687 | |||
| GenBank mRNA | AB051516.1 | - | 1 | 1979 | ||
| AK058072.2 | - | 1 | 818 | |||
| AK122755.1 | - | 1 | 1314 | |||
| AL833306.1 | - | 1 | 2163 | |||

ZNF518B
Genomic Map