siRNA Details
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053398.1 | NP_445850.1 | 8 | 1759 | ||
GenBank mRNA | BC079378.1 | - | 8 | 1759 | 7110848 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053398.1 | NP_445850.1 | 8 | 1759 | ||
GenBank mRNA | BC079378.1 | - | 8 | 1759 | 7110848 |