siRNA Details
| Gene Symbol: PKDCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138370.2 | NP_612379.2 | 5, 6 | 1407 | ||
| GenBank mRNA | BC007901.2 | - | 3, 4 | 362 | ||
| BC062988.1 | - | 3, 4 | 362 | |||
| BC094697.1 | - | 5, 6 | 815 | 30529646 | ||
| BC110513.1 | - | 5, 6 | 844 | 40031755 | ||
| BC110514.1 | - | 5, 6 | 844 | 40031756 | ||

PKDCC
Genomic Map