siRNA Details
| Gene Symbol: Adam34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145745.2 | NP_665688.2 | 2 | 2379 | ||
| GenBank mRNA | AF373288.1 | - | 3 | 2463 | ||
| AK029590.1 | - | 3 | 2432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam34| Gene Symbol: Adam34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145745.2 | NP_665688.2 | 2 | 2379 | ||
| GenBank mRNA | AF373288.1 | - | 3 | 2463 | ||
| AK029590.1 | - | 3 | 2432 | |||