siRNA Details
| Gene Symbol: Adam34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145745.2 | NP_665688.2 | 2 | 1870 | ||
| GenBank mRNA | AF373288.1 | - | 3 | 1954 | ||
| AK029590.1 | - | 3 | 1923 | |||
| BC132102.1 | - | 2 | 1824 | 40130379 | ||
| BC132292.1 | - | 2 | 1824 | 40130569 | ||
| BC141203.1 | - | 3 | 1913 | 9055815 | ||
| BC141204.1 | - | 3 | 1916 | 9055816 | ||

Adam34
Genomic Map