siRNA Details
| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198505.2 | NP_940907.2 | 3 | 347 | ||
| XM_011512770.1 | XP_011511072.1 | 3 | 446 | |||
| GenBank mRNA | AK122613.1 | - | 467 | |||
| AY823163.1 | - | 3 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP13A5| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198505.2 | NP_940907.2 | 3 | 347 | ||
| XM_011512770.1 | XP_011511072.1 | 3 | 446 | |||
| GenBank mRNA | AK122613.1 | - | 467 | |||
| AY823163.1 | - | 3 | 347 | |||