siRNA Details
| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198505.2 | NP_940907.2 | 30 | 3501 | ||
| XM_017006304.1 | XP_016861793.1 | 26 | 2814 | |||
| XM_017006305.1 | XP_016861794.1 | 17 | 1917 | |||
| GenBank mRNA | AY358667.1 | - | 4 | 310 | ||
| AY823163.1 | - | 30 | 3501 | |||

ATP13A5
Genomic Map