siRNA Details
| Gene Symbol: HTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000200.2 | NP_000191.1 | 6 | 321 | ||
| GenBank mRNA | BC009791.1 | - | 6 | 279 | 4293405 | |
| BC095438.1 | - | 6 | 278 | 30352697 | ||
| BP332860.1 | - | 6 | 275 | |||
| CB957931.1 | - | 6 | 321 | |||
| M18372.1 | - | 5 | 241 | |||
| M26665.1 | - | 6 | 275 | |||

HTN3
Genomic Map