siRNA Details
| Gene Symbol: Olr1240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000448.1 | NP_001000448.1 | 1 | 913 | ||
| Gene Symbol: LOC103692019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762664.1 | XP_008760886.1 | 1 | 913 | ||

Olr1240
Genomic Map