siRNA Details
| Gene Symbol: ASAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017707.3 | NP_060177.2 | 5 | 588 | ||
| XM_011541755.1 | XP_011540057.1 | 5 | 522 | |||
| XM_017001685.1 | XP_016857174.1 | 5 | 522 | |||
| XM_017001686.1 | XP_016857175.1 | 5 | 522 | |||
| XM_017001687.1 | XP_016857176.1 | 5 | 495 | |||
| XM_017001688.1 | XP_016857177.1 | 5 | 484 | |||
| XM_017001689.1 | XP_016857178.1 | 5 | 539 | |||
| GenBank mRNA | AB056749.1 | - | 5 | 509 | ||
| AK092291.1 | - | 5 | 510 | |||
| AK299359.1 | - | 5 | 484 | |||
| BC023519.2 | - | 6 | 576 | |||
| BC060786.1 | - | 5 | 536 | 5298355 | ||

ASAP3
Genomic Map