siRNA Details
| Gene Symbol: Olr1239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000811.1 | NP_001000811.1 | 1 | 492 | ||
| Gene Symbol: LOC103692040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762674.1 | XP_008760896.1 | 2 | 510 | ||

Olr1239
Genomic Map