siRNA Details
| Gene Symbol: Olr1199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000819.1 | NP_001000819.1 | 1 | 909 | ||
| Gene Symbol: LOC102551757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242799.1 | XP_006242861.1 | 1 | 507 | ||

Olr1199
Genomic Map