siRNA Details
| Gene Symbol: Olr1237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000963.1 | NP_001000963.1 | 1 | 522 | ||
| Gene Symbol: LOC100911939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749657.1 | XP_003749705.1 | 1 | 522 | ||

Olr1237
Genomic Map