siRNA Details
| Gene Symbol: Olr1238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001013.1 | NP_001001013.1 | 1 | 890 | ||
| Gene Symbol: LOC103692039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762673.1 | XP_008760895.1 | 1 | 890 | ||

Olr1238
Genomic Map