siRNA Details
| Gene Symbol: TMEM214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083590.1 | NP_001077059.1 | 16 | 2891 | ||
| NM_017727.4 | NP_060197.4 | 17 | 3026 | |||
| XM_005264381.3 | XP_005264438.1 | 18 | 3191 | |||
| XM_005264382.3 | XP_005264439.1 | 18 | 3077 | |||
| XM_005264383.3 | XP_005264440.1 | 10 | 2074 | |||
| GenBank mRNA | AK074981.1 | - | 17 | 2954 | ||
| AK097308.1 | - | 16 | 2821 | |||
| AL833725.1 | - | 17 | 2886 | |||
| BC068479.1 | - | 18 | 2689 | 4814193 | ||

TMEM214
Genomic Map