siRNA Details
| Gene Symbol: Olr1737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001422.1 | NP_001001422.1 | 1 | 909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1737| Gene Symbol: Olr1737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001422.1 | NP_001001422.1 | 1 | 909 | ||