siRNA Details
| Gene Symbol: OR4C13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001955.2 | NP_001001955.2 | 1 | 696 | ||
| GenBank mRNA | BC137005.1 | - | 1 | 696 | 9020998 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C13| Gene Symbol: OR4C13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001955.2 | NP_001001955.2 | 1 | 696 | ||
| GenBank mRNA | BC137005.1 | - | 1 | 696 | 9020998 | |