siRNA Details
| Gene Symbol: Tmem158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002267.2 | NP_001002267.2 | 1 | 560 | ||
| GenBank mRNA | AB114272.1 | - | 1 | 316 | ||
| AK009669.1 | - | 1 | 536 | |||
| BC139427.1 | - | 1 | 336 | 8862449 | ||
| BC139428.1 | - | 1 | 336 | 8862450 | ||

Tmem158
Genomic Map