siRNA Details
| Gene Symbol: Smg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002764.1 | NP_001002764.1 | 10 | 2850 | ||
| GenBank mRNA | AK129203.1 | - | 9 | 1469 | ||
| BC006644.1 | - | 2 | 165 | |||
| BC037179.1 | - | 8 | 1140 | |||
| BC047279.1 | - | 10 | 2862 | |||
| BC066040.1 | - | 10 | 2850 | 6410245 | ||
| BC077715.1 | - | 10 | 2813 | |||

Smg6
Genomic Map