siRNA Details
Gene Symbol: Prm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002855.2 | NP_001002855.1 | 1 | 200 | ||
GenBank mRNA | S79939.1 | - | 1 | 200 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002855.2 | NP_001002855.1 | 1 | 200 | ||
GenBank mRNA | S79939.1 | - | 1 | 200 |