siRNA Details
| Gene Symbol: AI413582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002895.2 | NP_001002895.2 | 5 | 451 | ||
| GenBank mRNA | AK131593.1 | - | 5 | 446 | ||
| AK131681.1 | - | 5 | 426 | |||
| BC028464.1 | - | 5 | 424 | 1264774 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI413582| Gene Symbol: AI413582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002895.2 | NP_001002895.2 | 5 | 451 | ||
| GenBank mRNA | AK131593.1 | - | 5 | 446 | ||
| AK131681.1 | - | 5 | 426 | |||
| BC028464.1 | - | 5 | 424 | 1264774 | ||