siRNA Details
| Gene Symbol: Higd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002900.1 | NP_001002900.1 | 1 | 35 | ||
| GenBank mRNA | AB099517.1 | - | 1 | 35 | ||
| AB253942.1 | - | 1 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Higd1c| Gene Symbol: Higd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002900.1 | NP_001002900.1 | 1 | 35 | ||
| GenBank mRNA | AB099517.1 | - | 1 | 35 | ||
| AB253942.1 | - | 1 | 75 | |||