siRNA Details
| Gene Symbol: LY6G6F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003693.1 | NP_001003693.1 | 3 | 460 | ||
| GenBank mRNA | AJ496460.1 | - | 3 | 460 | ||
| BC137212.1 | - | 3 | 504 | 9021209 | ||
| BC137213.1 | - | 3 | 504 | 9021210 | ||

LY6G6F
Genomic Map