siRNA Details
| Gene Symbol: TRIM6-TRIM34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003819.3 | NP_001003819.1 | 7, 8 | 1246 | ||
| GenBank mRNA | AB039903.1 | - | 7, 8 | 1154 | ||
| BC136871.1 | - | 7, 8 | 1165 | 9020862 | ||
| BC140722.1 | - | 7, 8 | 1165 | 9021590 | ||
| BC144424.1 | - | 7, 8 | 1165 | |||

TRIM6-TRIM34
Genomic Map