siRNA Details
Gene Symbol: HBM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003938.3 | NP_001003938.1 | 3 | 498 | ||
GenBank mRNA | AY698022.2 | - | 3 | 498 | ||
BC035682.1 | - | 3 | 481 | 5749846 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HBM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003938.3 | NP_001003938.1 | 3 | 498 | ||
GenBank mRNA | AY698022.2 | - | 3 | 498 | ||
BC035682.1 | - | 3 | 481 | 5749846 |