siRNA Details
| Gene Symbol: PLPPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017753.2 | NP_060223.2 | 8 | 2079 | ||
| NM_207299.1 | NP_997182.1 | 8 | 2212 | |||
| GenBank mRNA | AK000307.1 | - | 8 | 2077 | ||
| AY304515.1 | - | 8 | 1949 | |||
| BC022465.1 | - | 8 | 1949 | 4797229 | ||

PLPPR1
Genomic Map