siRNA Details
| Gene Symbol: Pisd-ps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034517.1 | - | 7 | 2981 | ||
| AK036319.1 | - | 7 | 2606 | |||
| AK144226.1 | - | 3 | 2320 | |||
| AK161130.1 | - | 7 | 2370 | |||
| AK162734.1 | - | 6 | 3721 | |||
| AK169597.1 | - | 4 | 3571 | |||
| AK169755.1 | - | 4 | 3571 | |||
| BC003217.1 | - | 1 | 1441 | |||
| BC034366.1 | - | 2 | 3340 | |||
| BC048401.1 | - | 5 | 2235 | |||
| BC059938.1 | - | 7 | 1664 | |||
| BC083061.1 | - | 6 | 2072 | |||

Pisd-ps3
Genomic Map