siRNA Details
| Gene Symbol: Pisd-ps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005333.1 | - | 440 | |||
| AK017116.1 | - | 2 | 345 | |||
| AK034517.1 | - | 2 | 993 | |||
| AK036319.1 | - | 2 | 266 | |||
| AK081907.1 | - | 2 | 313 | |||
| AK161130.1 | - | 2 | 336 | |||
| AK162734.1 | - | 2 | 1469 | |||
| AK168892.1 | - | 2 | 313 | |||
| AK169597.1 | - | 1 | 1079 | |||
| AK169755.1 | - | 1 | 1079 | |||
| AK185609.1 | - | 1 | 248 | |||
| AK185737.1 | - | 1 | 254 | |||
| AK199714.1 | - | 1 | 246 | |||
| BC048401.1 | - | 1 | 366 | |||
| BC059938.1 | - | 1 | 242 | |||
| BC083061.1 | - | 1 | 84 | |||
| Gene Symbol: Pisd-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG076997.2 | - | 2 | 331 | ||
| CX562509.1 | - | 1 | 194 | |||

Pisd-ps3
Genomic Map