siRNA Details
| Gene Symbol: Rragb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004154.2 | NP_001004154.1 | 7 | 1171 | ||
| GenBank mRNA | BC069180.1 | - | 7 | 1129 | 6810543 | |
| BC070407.1 | - | 7 | 1148 | 6808979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rragb| Gene Symbol: Rragb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004154.2 | NP_001004154.1 | 7 | 1171 | ||
| GenBank mRNA | BC069180.1 | - | 7 | 1129 | 6810543 | |
| BC070407.1 | - | 7 | 1148 | 6808979 | ||