siRNA Details
| Gene Symbol: Gm1553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001255990.1 | NP_001242919.1 | 4 | 374 | ||
| GenBank mRNA | AY702028.1 | - | 4 | 374 | ||
| BC141250.1 | - | 4 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1553| Gene Symbol: Gm1553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001255990.1 | NP_001242919.1 | 4 | 374 | ||
| GenBank mRNA | AY702028.1 | - | 4 | 374 | ||
| BC141250.1 | - | 4 | 364 | |||