siRNA Details
| Gene Symbol: LINC01002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028324.1 | 3 | 602 | |||
| GenBank mRNA | AK128780.1 | - | 1 | 141 | ||
| AK303004.1 | - | 3 | 566 | |||
| Gene Symbol: LINC01000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024368.1 | 5 | 828 | |||
| GenBank mRNA | AK302487.1 | - | 5 | 834 | ||
| AK304049.1 | - | 5 | 808 | |||
| BC062355.1 | - | 5 | 732 | |||
| BC131589.1 | - | 5 | 732 | |||
| Gene Symbol: LINC01001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028326.1 | 3 | 602 | |||
| GenBank mRNA | AK293878.1 | - | 3 | 630 | ||
| Gene Symbol: LOC100132062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028325.1 | 3 | 602 | |||
| Gene Symbol: LOC100132287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028322.1 | 3 | 602 | |||
| Gene Symbol: LOC100133331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028327.1 | 3 | 602 | |||
| Gene Symbol: LOC101929819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK302511.1 | - | 7 | 1082 | ||

LINC01002
Genomic Map