siRNA Details
| Gene Symbol: OR10A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004460.1 | NP_001004460.1 | 1 | 60 | ||
| GenBank mRNA | BC136955.1 | - | 1 | 122 | 9020947 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10A2| Gene Symbol: OR10A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004460.1 | NP_001004460.1 | 1 | 60 | ||
| GenBank mRNA | BC136955.1 | - | 1 | 122 | 9020947 | |