siRNA Details
| Gene Symbol: OR2AG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004490.1 | NP_001004490.1 | 1 | 527 | ||
| GenBank mRNA | BC140734.1 | - | 1 | 625 | 9021602 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2AG2| Gene Symbol: OR2AG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004490.1 | NP_001004490.1 | 1 | 527 | ||
| GenBank mRNA | BC140734.1 | - | 1 | 625 | 9021602 | |