siRNA Details
Gene Symbol: OR2L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002145.1 | 1 | 759 |
Gene Symbol: OR2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004686.2 | NP_001004686.1 | 1 | 854 | ||
XM_011544158.2 | XP_011542460.1 | 2 | 984 | |||
GenBank mRNA | BC104792.1 | - | 1 | 854 | 8143795 | |
BC104794.1 | - | 1 | 854 | 8143797 |
Gene Symbol: OR2L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258284.1 | NP_001245213.1 | 1 | 758 |
