siRNA Details
| Gene Symbol: OR2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004686.2 | NP_001004686.1 | 1 | 736 | ||
| XM_011544158.2 | XP_011542460.1 | 2 | 866 | |||
| GenBank mRNA | BC104792.1 | - | 1 | 736 | 8143795 | |
| BC104794.1 | - | 1 | 736 | 8143797 | ||
| X64978.1 | - | 1 | 269 | |||

OR2L2
Genomic Map