siRNA Details
| Gene Symbol: OR52I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005170.2 | NP_001005170.1 | 1 | 1041 | ||
| GenBank mRNA | BC136921.1 | - | 1 | 1041 | 9020912 | |
| BC151144.1 | - | 1 | 1041 | 9020913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52I2| Gene Symbol: OR52I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005170.2 | NP_001005170.1 | 1 | 1041 | ||
| GenBank mRNA | BC136921.1 | - | 1 | 1041 | 9020912 | |
| BC151144.1 | - | 1 | 1041 | 9020913 | ||