siRNA Details
| Gene Symbol: OR8G5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005198.1 | NP_001005198.1 | 1 | 574 | ||
| XM_011542662.2 | XP_011540964.1 | 4 | 1128 | |||
| XM_011542663.2 | XP_011540965.1 | 4 | 898 | |||
| XM_011542664.2 | XP_011540966.1 | 3 | 767 | |||
| XM_011542665.2 | XP_011540967.1 | 3 | 1048 | |||
| XM_011542666.2 | XP_011540968.1 | 3 | 818 | |||
| XM_011542667.2 | XP_011540969.1 | 2 | 687 | |||
| GenBank mRNA | BC136820.1 | - | 1 | 597 | 9020811 | |
| BC136822.1 | - | 1 | 597 | 9020813 | ||

OR8G5
Genomic Map