siRNA Details
Gene Symbol: OR4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005272.3 | NP_001005272.3 | 1 | 567 | ||
GenBank mRNA | BC140750.1 | - | 1 | 567 | 9021618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005272.3 | NP_001005272.3 | 1 | 567 | ||
GenBank mRNA | BC140750.1 | - | 1 | 567 | 9021618 |