siRNA Details
| Gene Symbol: G2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308097.1 | NP_001295026.1 | 2 | 220 | ||
| NM_017769.4 | NP_060239.2 | 3 | 261 | |||
| XM_011536925.2 | XP_011535227.1 | 3 | 227 | |||
| XM_011536926.1 | XP_011535228.1 | 3 | 261 | |||
| GenBank mRNA | AB037754.1 | - | 3 | 213 | ||
| AK000340.1 | - | 3 | 226 | |||
| AK093536.1 | - | 2 | 185 | |||
| AK225346.1 | - | 3 | 226 | |||
| AK295571.1 | - | 2 | 185 | |||
| AK296872.1 | - | 2 | 185 | |||
| BC000973.2 | - | 3 | 207 | 3450488 | ||
| DA491192.1 | - | 3 | 261 | |||

G2E3
Genomic Map